Assay Details
| Gene Symbol: Nfya | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846610.3 | XP_004846667.1 | 8 - 9 | 1130 | 60 | |
| XM_004846611.3 | XP_004846668.1 | 8 - 9 | 2081 | 60 | ||
| XM_004846612.3 | XP_004846669.1 | 8 - 9 | 1127 | 60 | ||
| XM_004846613.3 | XP_004846670.1 | 8 - 9 | 1112 | 60 | ||
| XM_013072841.2 | XP_012928295.1 | 7 - 8 | 1043 | 60 | ||
| XM_013072842.2 | XP_012928296.1 | 7 - 8 | 1025 | 60 | ||
| XM_021259861.1 | XP_021115520.1 | 9 - 10 | 3126 | 60 | ||
| XM_021259862.1 | XP_021115521.1 | 8 - 9 | 1127 | 60 | ||
| XM_021259864.1 | XP_021115523.1 | 8 - 9 | 2078 | 60 | ||
| XM_021259865.1 | XP_021115524.1 | 8 - 9 | 1124 | 60 | ||
| XM_021259866.1 | XP_021115525.1 | 7 - 8 | 1992 | 60 | ||
More Information
Set Membership: |
|

Nfya
Genomic Map