Assay Details
Gene Symbol: LOC101719334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004843641.2 | XP_004843698.1 | 4 - 5 | 1041 | 90 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
