Assay Details
| Gene Symbol: Col21a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013067094.2 | XP_012922548.1 | 10 - 11 | 2043 | 62 | |
| XM_021247614.1 | XP_021103273.1 | 10 - 11 | 2044 | 62 | ||
More Information
Set Membership: |
|

Col21a1
Genomic Map