Assay Details
| Gene Symbol: Gne | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870102.3 | XP_004870159.1 | 11 - 12 | 2154 | 57 | |
| XM_004870104.3 | XP_004870161.1 | 11 - 12 | 2150 | 57 | ||
| XM_004870105.3 | XP_004870162.1 | 11 - 12 | 2243 | 57 | ||
| XM_004870106.1 | XP_004870163.1 | 10 - 11 | 1921 | 57 | ||
| XM_004870107.3 | XP_004870164.1 | 11 - 12 | 2094 | 57 | ||
| XM_004870108.3 | XP_004870165.1 | 11 - 12 | 2144 | 57 | ||
| XM_021248505.1 | XP_021104164.1 | 11 - 12 | 1992 | 57 | ||
| XM_021248506.1 | XP_021104165.1 | 10 - 11 | 2037 | 57 | ||
| XM_021248507.1 | XP_021104166.1 | 10 - 11 | 2043 | 57 | ||
| XM_021248508.1 | XP_021104167.1 | 10 - 11 | 1791 | 57 | ||
More Information
Set Membership: |
|

Gne
Genomic Map