Assay Details
| Gene Symbol: Fst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848529.3 | XP_004848586.1 | 3 - 4 | 904 | 59 | |
| XM_004848530.3 | XP_004848587.1 | 3 - 4 | 905 | 59 | ||
| XM_004848531.3 | XP_004848588.1 | 3 - 4 | 905 | 59 | ||
More Information
Set Membership: |
|

Fst
Genomic Map