Assay Details
| Gene Symbol: Trpv6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860515.3 | XP_004860572.1 | 3 - 4 | 625 | 74 | |
| XM_013078011.2 | XP_012933465.1 | 3 - 4 | 641 | 74 | ||
| XM_021240932.1 | XP_021096591.1 | 3 - 4 | 626 | 74 | ||
| XM_021240933.1 | XP_021096592.1 | 3 - 4 | 626 | 74 | ||
More Information
Set Membership: |
|

Trpv6
Genomic Map