Assay Details
| Gene Symbol: Rpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851118.2 | XP_004851175.1 | 2 - 3 | 339 | 65 | |
| XM_004851119.2 | XP_004851176.1 | 2 - 3 | 339 | 65 | ||
| XM_004851120.2 | XP_004851177.1 | 2 - 3 | 339 | 65 | ||
| XM_021263501.1 | XP_021119160.1 | 2 - 3 | 338 | 65 | ||
| XM_021263502.1 | XP_021119161.1 | 2 - 3 | 339 | 65 | ||
| XM_021263503.1 | XP_021119162.1 | 2 - 3 | 339 | 65 | ||
| XM_021263505.1 | XP_021119164.1 | 2 - 3 | 339 | 65 | ||
| XM_021263506.1 | XP_021119165.1 | 2 - 3 | 339 | 65 | ||
More Information
Set Membership: |
|

Rpe
Genomic Map