Assay Details
| Gene Symbol: LOC101707009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852778.2 | XP_004852835.1 | 4 - 5 | 483 | 124 | |
| XM_021264822.1 | XP_021120481.1 | 4 - 5 | 484 | 124 | ||
More Information
Set Membership: |
|

LOC101707009
Genomic Map