Assay Details
| Gene Symbol: LOC101707009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852778.2 | XP_004852835.1 | 5 - 6 | 589 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101707009| Gene Symbol: LOC101707009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852778.2 | XP_004852835.1 | 5 - 6 | 589 | 94 | |
Set Membership: |
|