Assay Details
| Gene Symbol: Mapk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836741.3 | XP_004836798.1 | 9 - 10 | 1187 | 61 | |
| XM_004836742.3 | XP_004836799.1 | 9 - 10 | 1190 | 61 | ||
| XM_004836746.3 | XP_004836803.1 | 9 - 10 | 1190 | 61 | ||
| XM_021242594.1 | XP_021098253.1 | 9 - 10 | 1276 | 61 | ||
| XM_021242605.1 | XP_021098264.1 | 7 - 8 | 984 | 61 | ||
More Information
Set Membership: |
|

Mapk9
Genomic Map