Assay Details
| Gene Symbol: Nmnat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834343.2 | XP_004834400.1 | 7 - 8 | 961 | 62 | |
| XM_004834344.2 | XP_004834401.1 | 4 - 5 | 643 | 62 | ||
| XM_013070906.2 | XP_012926360.1 | 6 - 7 | 898 | 62 | ||
| XM_013070911.2 | XP_012926365.1 | 6 - 7 | 863 | 62 | ||
| XM_013070920.2 | XP_012926374.1 | 3 - 4 | 580 | 62 | ||
| XM_021250364.1 | XP_021106023.1 | 5 - 6 | 869 | 62 | ||
| XM_021250443.1 | XP_021106102.1 | 9 - 10 | 1292 | 62 | ||
| XM_021250600.1 | XP_021106259.1 | 3 - 4 | 437 | 62 | ||
More Information
Set Membership: |
|

Nmnat3
Genomic Map