Assay Details
| Gene Symbol: Hyal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834130.3 | XP_004834187.1 | 2 - 3 | 1069 | 74 | |
| XM_004834131.3 | XP_004834188.1 | 2 - 3 | 1153 | 74 | ||
More Information
Set Membership: |
|

Hyal2
Genomic Map