Assay Details
| Gene Symbol: Sdhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856577.3 | XP_004856634.1 | 1 - 2 | 166 | 60 | |
| XM_004856579.3 | XP_004856636.1 | 1 - 2 | 166 | 60 | ||
More Information
Set Membership: |
|

Sdhd
Genomic Map