Assay Details
| Gene Symbol: LOC101703023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013067201.2 | XP_012922655.1 | 4 - 5 | 746 | 70 | |
| XM_021247831.1 | XP_021103490.1 | 5 - 6 | 736 | 70 | ||
More Information
Set Membership: |
|

LOC101703023
Genomic Map