Assay Details
| Gene Symbol: LOC101706975 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866656.3 | XP_004866713.1 | 2 - 3 | 261 | 63 | |
| XM_004866657.3 | XP_004866714.1 | 2 - 3 | 269 | 63 | ||
More Information
Set Membership: |
|

LOC101706975
Genomic Map