Assay Details
| Gene Symbol: LOC101712592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013072957.2 | XP_012928411.1 | 5 - 6 | 895 | 71 | |
| XM_021260181.1 | XP_021115840.1 | 5 - 6 | 1418 | 71 | ||
More Information
Set Membership: |
|

LOC101712592
Genomic Map