Assay Details
| Gene Symbol: Trove2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862763.3 | XP_004862820.1 | 3 - 4 | 964 | 85 | |
| XM_021242910.1 | XP_021098569.1 | 3 - 4 | 964 | 85 | ||
More Information
Set Membership: |
|

Trove2
Genomic Map