Assay Details
| Gene Symbol: Rbm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871937.3 | XP_004871994.1 | 4 - 5 | 484 | 59 | |
| XM_021249983.1 | XP_021105642.1 | 4 - 5 | 539 | 59 | ||
| XM_021249984.1 | XP_021105643.1 | 3 - 4 | 345 | 59 | ||
| XM_021249985.1 | XP_021105644.1 | 4 - 5 | 533 | 59 | ||
More Information
Set Membership: |
|

Rbm3
Genomic Map