Assay Details
| Gene Symbol: LOC101710152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021241506.1 | XP_021097165.1 | 1 - 2 | 208 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021241506.1 transcripts.
Set Membership: |
|

LOC101710152
Genomic Map