Assay Details
| Gene Symbol: LOC101710152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861305.3 | XP_004861362.1 | 2 - 3 | 398 | 68 | |
| XM_021241505.1 | XP_021097164.1 | 2 - 3 | 383 | 68 | ||
| XM_021241506.1 | XP_021097165.1 | 2 - 3 | 325 | 68 | ||
| XM_021241507.1 | XP_021097166.1 | 2 - 3 | 390 | 68 | ||
More Information
Set Membership: |
|

LOC101710152
Genomic Map