Assay Details
| Gene Symbol: LOC101710152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861305.3 | XP_004861362.1 | 4 - 5 | 713 | 79 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101710152| Gene Symbol: LOC101710152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861305.3 | XP_004861362.1 | 4 - 5 | 713 | 79 | |