Assay Details
| Gene Symbol: Znf148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835221.2 | XP_004835278.1 | 4 - 5 | 766 | 92 | |
| XM_004835222.3 | XP_004835279.2 | 5 - 6 | 878 | 92 | ||
| XM_013073893.2 | XP_012929347.1 | 4 - 5 | 766 | 92 | ||
| XM_021259642.1 | XP_021115301.1 | 3 - 4 | 500 | 92 | ||
| XM_021259646.1 | XP_021115305.1 | 4 - 5 | 582 | 92 | ||
| XM_021259655.1 | XP_021115314.1 | 4 - 5 | 766 | 92 | ||
More Information
Set Membership: |
|

Znf148
Genomic Map