Assay Details
| Gene Symbol: LOC101717096 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013068548.2 | XP_012924002.1 | 1 - 2 | 827 | 90 | |
| XM_021250725.1 | XP_021106384.1 | 1 - 2 | 829 | 90 | ||
More Information
Set Membership: |
|

LOC101717096
Genomic Map