Assay Details
| Gene Symbol: LOC101717096 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872947.3 | XP_004873004.1 | 5 - 6 | 748 | 102 | |
| XM_013068548.2 | XP_012924002.1 | 5 - 6 | 1341 | 102 | ||
| XM_021250724.1 | XP_021106383.1 | 5 - 6 | 585 | 102 | ||
| XM_021250725.1 | XP_021106384.1 | 5 - 6 | 1343 | 102 | ||
More Information
Set Membership: |
|

LOC101717096
Genomic Map