Assay Details
| Gene Symbol: Chmp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857906.3 | XP_004857963.1 | 3 - 4 | 590 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chmp2b| Gene Symbol: Chmp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857906.3 | XP_004857963.1 | 3 - 4 | 590 | 71 | |
Set Membership: |
|