Assay Details
| Gene Symbol: LOC101719875 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856628.3 | XP_004856685.1 | 4 - 5 | 1996 | 71 | |
| XM_021237808.1 | XP_021093467.1 | 3 - 4 | 2023 | 71 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101719875| Gene Symbol: LOC101719875 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856628.3 | XP_004856685.1 | 4 - 5 | 1996 | 71 | |
| XM_021237808.1 | XP_021093467.1 | 3 - 4 | 2023 | 71 | ||