Assay Details
| Gene Symbol: Naga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021258900.1 | XP_021114559.1 | 3 - 4 | 331 | 59 | |
| XM_021258901.1 | XP_021114560.1 | 3 - 4 | 328 | 59 | ||
| XM_021258902.1 | XP_021114561.1 | 3 - 4 | 328 | 59 | ||
| XM_021258903.1 | XP_021114562.1 | 3 - 4 | 328 | 59 | ||
| XM_021258904.1 | XP_021114563.1 | 3 - 4 | 328 | 59 | ||
| XM_021258905.1 | XP_021114564.1 | 3 - 4 | 328 | 59 | ||
More Information
Set Membership: |
|

Naga
Genomic Map