Assay Details
| Gene Symbol: Lrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858918.3 | XP_004858975.1 | 6 - 7 | 1789 | 65 | |
| XM_013077535.2 | XP_012932989.1 | 4 - 5 | 2644 | 65 | ||
More Information
Set Membership: |
|

Lrp3
Genomic Map