Assay Details
| Gene Symbol: Brca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854764.2 | XP_004854821.1 | 18 - 19 | 8601 | 77 | |
| XM_021266507.1 | XP_021122166.1 | 18 - 19 | 8580 | 77 | ||
| XM_021266508.1 | XP_021122167.1 | 18 - 19 | 8523 | 77 | ||
| XM_021266509.1 | XP_021122168.1 | 14 - 15 | 7999 | 77 | ||
| XM_021266510.1 | XP_021122169.1 | 18 - 19 | 8601 | 77 | ||
| XM_021266511.1 | XP_021122170.1 | 18 - 19 | 8601 | 77 | ||
More Information
Set Membership: |
|

Brca2
Genomic Map