Assay Details
| Gene Symbol: Mre11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838047.2 | XP_004838104.1 | 18 - 19 | 2241 | 94 | |
| XM_021252663.1 | XP_021108322.1 | 18 - 19 | 2192 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mre11| Gene Symbol: Mre11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838047.2 | XP_004838104.1 | 18 - 19 | 2241 | 94 | |
| XM_021252663.1 | XP_021108322.1 | 18 - 19 | 2192 | 94 | ||