Assay Details
| Gene Symbol: Mre11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838047.2 | XP_004838104.1 | 6 - 7 | 790 | 79 | |
| XM_013069640.2 | XP_012925094.1 | 6 - 7 | 790 | 79 | ||
| XM_021252663.1 | XP_021108322.1 | 6 - 7 | 741 | 79 | ||

Mre11
Genomic Map