Assay Details
| Gene Symbol: Bbx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835766.3 | XP_004835823.1 | 17 - 18 | 3037 | 59 | |
| XM_004835767.3 | XP_004835824.1 | 18 - 19 | 3154 | 59 | ||
| XM_004835768.3 | XP_004835825.1 | 17 - 18 | 3042 | 59 | ||
| XM_004835769.3 | XP_004835826.1 | 16 - 17 | 2964 | 59 | ||
| XM_004835770.3 | XP_004835827.1 | 16 - 17 | 3062 | 59 | ||
| XM_021264144.1 | XP_021119803.1 | 15 - 16 | 2789 | 59 | ||
| XM_021264145.1 | XP_021119804.1 | 14 - 15 | 2680 | 59 | ||
| XM_021264156.1 | XP_021119815.1 | 17 - 18 | 2994 | 59 | ||
| XM_021264165.1 | XP_021119824.1 | 17 - 18 | 2912 | 59 | ||
| XM_021264174.1 | XP_021119833.1 | 15 - 16 | 2839 | 59 | ||
| XM_021264181.1 | XP_021119840.1 | 16 - 17 | 2966 | 59 | ||
| XM_021264192.1 | XP_021119851.1 | 16 - 17 | 2842 | 59 | ||
| XM_021264210.1 | XP_021119869.1 | 15 - 16 | 2945 | 59 | ||
| XM_021264214.1 | XP_021119873.1 | 14 - 15 | 2762 | 59 | ||
| XM_021264215.1 | XP_021119874.1 | 16 - 17 | 2921 | 59 | ||
More Information
Set Membership: |
|

Bbx
Genomic Map