Assay Details
| Gene Symbol: Msmo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848223.2 | XP_004848280.1 | 5 - 6 | 850 | 91 | |
| XM_004848224.3 | XP_004848281.1 | 5 - 6 | 747 | 91 | ||
More Information
Set Membership: |
|

Msmo1
Genomic Map