Assay Details
| Gene Symbol: Rps15a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855795.2 | XP_004855852.1 | 1 - 2 | 93 | 63 | |
| XM_021237225.1 | XP_021092884.1 | 1 - 2 | 95 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004855795.2, XM_021237225.1 transcripts.
Set Membership: |
|

Rps15a
Genomic Map