Assay Details
| Gene Symbol: Rps19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873069.3 | XP_004873126.1 | 3 - 4 | 369 | 72 | |
| XM_021250981.1 | XP_021106640.1 | 3 - 4 | 195 | 72 | ||
More Information
Set Membership: |
|

Rps19
Genomic Map