Assay Details
| Gene Symbol: Rps17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852847.2 | XP_004852904.1 | 1 - 2 | 231 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004852847.2 transcripts.
Set Membership: |
|

Rps17
Genomic Map