Assay Details
| Gene Symbol: Rps8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872902.2 | XP_004872959.1 | 1 - 2 | 101 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004872902.2 transcripts.
Set Membership: |
|

Rps8
Genomic Map