Assay Details
| Gene Symbol: Serpinc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853407.3 | XP_004853464.1 | 7 - 8 | 1498 | 58 | |
| XM_004853409.3 | XP_004853466.1 | 7 - 8 | 1449 | 58 | ||
| XM_004853412.3 | XP_004853469.1 | 6 - 7 | 1365 | 58 | ||
| XM_021265400.1 | XP_021121059.1 | 7 - 8 | 1592 | 58 | ||
More Information
Set Membership: |
|

Serpinc1
Genomic Map