Assay Details
| Gene Symbol: Cstb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842499.3 | XP_004842556.1 | 1 - 2 | 185 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004842499.3 transcripts.
Set Membership: |
|

Cstb
Genomic Map