Assay Details
| Gene Symbol: Sdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013074141.2 | XP_012929595.1 | 3 - 4 | 813 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sdc2| Gene Symbol: Sdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013074141.2 | XP_012929595.1 | 3 - 4 | 813 | 83 | |
Set Membership: |
|