Assay Details
| Gene Symbol: Suclg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844630.3 | XP_004844687.2 | 5 - 6 | 633 | 61 | |
| XM_021258242.1 | XP_021113901.1 | 5 - 6 | 520 | 61 | ||
More Information
Set Membership: |
|

Suclg1
Genomic Map