Assay Details
| Gene Symbol: Pea15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858725.3 | XP_004858782.1 | 1 - 2 | 154 | 96 | |
| XM_013077481.2 | XP_012932935.1 | 1 - 2 | 154 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004858725.3, XM_013077481.2 transcripts.
Set Membership: |
|

Pea15
Genomic Map