Assay Details
| Gene Symbol: Pea15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858725.3 | XP_004858782.1 | 2 - 3 | 328 | 76 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pea15| Gene Symbol: Pea15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858725.3 | XP_004858782.1 | 2 - 3 | 328 | 76 | |