Assay Details
| Gene Symbol: Prkar2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839671.3 | XP_004839728.1 | 7 - 8 | 933 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkar2b| Gene Symbol: Prkar2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839671.3 | XP_004839728.1 | 7 - 8 | 933 | 83 | |
Set Membership: |
|