Assay Details
| Gene Symbol: Fam96a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855669.3 | XP_004855726.1 | 2 - 3 | 468 | 73 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam96a| Gene Symbol: Fam96a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855669.3 | XP_004855726.1 | 2 - 3 | 468 | 73 | |