Assay Details
| Gene Symbol: Tsg101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851536.3 | XP_004851593.1 | 1 - 2 | 162 | 67 | |
| XM_021263905.1 | XP_021119564.1 | 1 - 2 | 162 | 67 | ||
More Information
Set Membership: |
|

Tsg101
Genomic Map