Assay Details
| Gene Symbol: Pycr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021242245.1 | XP_021097904.1 | 2 - 3 | 306 | 70 | |
| XM_021242246.1 | XP_021097905.1 | 2 - 3 | 306 | 70 | ||
| XM_021242247.1 | XP_021097906.1 | 2 - 3 | 306 | 70 | ||
| XM_021242248.1 | XP_021097907.1 | 2 - 3 | 306 | 70 | ||
| XM_021242249.1 | XP_021097908.1 | 2 - 3 | 306 | 70 | ||
| XM_021242250.1 | XP_021097909.1 | 2 - 3 | 308 | 70 | ||
| XM_021242251.1 | XP_021097910.1 | 2 - 3 | 269 | 70 | ||
More Information
Set Membership: |
|

Pycr2
Genomic Map