Assay Details
| Gene Symbol: Magohb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845891.2 | XP_004845948.1 | 3 - 4 | 363 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Magohb| Gene Symbol: Magohb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845891.2 | XP_004845948.1 | 3 - 4 | 363 | 94 | |
Set Membership: |
|