Assay Details
| Gene Symbol: Ahcy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867918.3 | XP_004867975.1 | 5 - 6 | 739 | 60 | |
| XM_004867919.3 | XP_004867976.1 | 5 - 6 | 739 | 60 | ||
| XM_004867920.3 | XP_004867977.1 | 5 - 6 | 788 | 60 | ||
| XM_021246884.1 | XP_021102543.1 | 5 - 6 | 739 | 60 | ||
More Information
Set Membership: |
|

Ahcy
Genomic Map