Assay Details
| Gene Symbol: Fabp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842005.3 | XP_004842062.1 | 3 - 4 | 420 | 85 | |
| XM_021255989.1 | XP_021111648.1 | 2 - 3 | 232 | 85 | ||
More Information
Set Membership: |
|

Fabp5
Genomic Map